Gene Transcripts

Gene Symbol
Kcng1
Entrez Gene ID
296395
Gene Name
potassium channel, voltage-gated modifier subfamily G, member 1
Gene Aliases
Kh2, Kv6.1
Chromosome Location
Chr. 3: 165020067 - 165039707 on Build Rnor_6.0
UniGene ID
Rn.91149
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001106545.1 NP_001100015.1 3 1609
XM_008762484.1 XP_008760706.1 3 1428
GenBank mRNA M81784.1 - 3 1434

View Full Product Details