Gene Transcripts

Gene Symbol
Cables2
Entrez Gene ID
311703
Gene Name
Cdk5 and Abl enzyme substrate 2
Gene Aliases
Chromosome Location
Chr. 3: 175630266 - 175644994 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_006235831.2 XP_006235893.2 3 511
XM_008762585.1 XP_008760807.1 3 511
XM_008762586.1 XP_008760808.1 3 511
XM_008762588.1 XP_008760810.1 2 439
XM_008762589.1 XP_008760811.1 3 511
XM_008762590.1 XP_008760812.1 3 511
XM_008762591.1 XP_008760813.1 3 511

View Full Product Details