Gene Transcripts

Gene Symbol
Fam46b
Entrez Gene ID
313019
Gene Name
family with sequence similarity 46, member B
Gene Aliases
RGD1308967
Chromosome Location
Chr. 5: 151692068 - 151699381 on Build Rnor_6.0
UniGene ID
Rn.143980
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001107907.2 NP_001101377.1 2 434
GenBank mRNA BC158863.1 - 2 434

Gene Symbol
LOC680624
Entrez Gene ID
680624
Gene Name
similar to CG30497-PA, isoform A
Gene Aliases
Chromosome Location
Chr. 11: 12275169 - 12277833 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008768543.1 XP_008766765.1 2 1075

View Full Product Details