Gene Transcripts

Gene Symbol
Ap3d1
Entrez Gene ID
314633
Gene Name
adaptor-related protein complex 3, delta 1 subunit
Gene Aliases
Ap3d
Chromosome Location
Chr. 7: 11838639 - 11873992 on Build Rnor_6.0
UniGene ID
Rn.8074
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100719.1 NP_001094189.1 11, 12 1183
GenBank mRNA BC169097.1 - 11, 12 1183

View Full Product Details