Gene Transcripts

Gene Symbol
Sned1
Entrez Gene ID
316638
Gene Name
sushi, nidogen and EGF-like domains 1
Gene Aliases
IRE-BP1
Chromosome Location
Chr. 9: 100380486 - 100439125 on Build Rnor_6.0
UniGene ID
Rn.23447
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001167842.1 NP_001161314.1 7 1209
XM_006245526.2 XP_006245588.1 7 1237
XM_008767373.1 XP_008765595.1 7 1238
XM_008767374.1 XP_008765596.1 7 1238

View Full Product Details