Gene Transcripts

Gene Symbol
Myh4
Entrez Gene ID
360543
Gene Name
myosin, heavy chain 4, skeletal muscle
Gene Aliases
Chromosome Location
Chr. 10: 53778456 - 53801605 on Build Rnor_6.0
UniGene ID
Rn.224464
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019325.1 NP_062198.1 30 4431
GenBank mRNA BC113948.1 - 30 4431
FQ224129.1 - 6 1286

View Full Product Details