Gene Transcripts

Gene Symbol
Gsdma
Entrez Gene ID
360619
Gene Name
gasdermin A
Gene Aliases
Gsdma1
Chromosome Location
Chr. 10: 86569903 - 86595102 on Build Rnor_6.0
UniGene ID
Rn.82083
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108297.1 NP_001101767.1 12 1223
XM_006247347.1 XP_006247409.1 12 1258
GenBank mRNA BC112385.1 - 11 2093

View Full Product Details