Gene Transcripts

Gene Symbol
Srcap
Entrez Gene ID
361652
Gene Name
Snf2-related CREBBP activator protein
Gene Aliases
RGD1565642
Chromosome Location
Chr. 1: 198957765 - 199007576 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008759936.1 XP_008758158.1 19 4251

View Full Product Details