Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001010964.1 | NP_001010964.1 | 3, 4 | 283 | |
XM_006237414.2 | XP_006237476.1 | 3, 4 | 343 | ||
GenBank mRNA | EF100677.1 | - | 3, 4 | 267 | |
EF100681.1 | - | 3, 4 | 267 | ||
EF100683.1 | - | 3, 4 | 267 | ||
FJ416340.1 | - | 3, 4 | 288 | ||
M62891.1 | - | 3, 4 | 283 |