Gene Transcripts

Gene Symbol
BPY2
Entrez Gene ID
9083
Gene Name
basic charge, Y-linked, 2
Gene Aliases
BPY2A, VCY2, VCY2A
Chromosome Location
Chr. Y: 22984263 - 23005465 on Build GRCh38
UniGene ID
Hs.664724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004678.2 NP_004669.2 6 605
GenBank mRNA AF000980.1 - 6 605

Gene Symbol
BPY2B
Entrez Gene ID
442867
Gene Name
basic charge, Y-linked, 2B
Gene Aliases
VCY2B
Chromosome Location
Chr. Y: 24618004 - 24639207 on Build GRCh38
UniGene ID
Hs.632848
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002760.1 NP_001002760.1 6 605

Gene Symbol
BPY2C
Entrez Gene ID
442868
Gene Name
basic charge, Y-linked, 2C
Gene Aliases
VCY2C
Chromosome Location
Chr. Y: 25030901 - 25052104 on Build GRCh38
UniGene ID
Hs.664724
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001002761.1 NP_001002761.1 6 605

View Full Product Details