Gene Transcripts

Gene Symbol
Olr86
Entrez Gene ID
405014
Gene Name
olfactory receptor 86
Gene Aliases
Chromosome Location
Chr. 1: 168378637 - 168379575 on Build Rnor_6.0
UniGene ID
Rn.141900
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001000747.1 NP_001000747.1 1 895

View Full Product Details