Gene Transcripts
- Gene Symbol
- Ly49i4
- Entrez Gene ID
- 494204
- Gene Name
- Ly49 inhibitory receptor 4
- Gene Aliases
- Chromosome Location
- Chr. 4: 164515105 - 164536581 on Build Rnor_6.0
- UniGene ID
- Rn.83631
- Species
- Rattus norvegicus
- Gene Symbol
- Ly49s6
- Entrez Gene ID
- 494196
- Gene Name
- Ly49 stimulatory receptor 6
- Gene Aliases
- Chromosome Location
- Chr. 4: 164429048 - 164453171 on Build Rnor_6.0
- UniGene ID
- Rn.126999
- Species
- Rattus norvegicus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC088257.1 |
- |
4 |
462 |
|
View Full Product Details