Gene Transcripts
- Gene Symbol
- CCPG1
- Entrez Gene ID
- 9236
- Gene Name
- cell cycle progression 1
- Gene Aliases
- CPR8
- Chromosome Location
- Chr. 15: 55355223 - 55408510 on Build GRCh38
- UniGene ID
- Hs.285051
- Species
- Homo sapiens
- Gene Symbol
- DYX1C1-CCPG1
- Entrez Gene ID
- 100533483
- Gene Name
- DYX1C1-CCPG1 readthrough (NMD candidate)
- Gene Aliases
- Chromosome Location
- Chr. 15: 55355223 - 55498584 on Build GRCh38
- UniGene ID
- Hs.285051
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037923.1 |
|
15 |
2400 |
|
View Full Product Details