Gene Transcripts

Gene Symbol
Mettl8
Entrez Gene ID
502633
Gene Name
methyltransferase like 8
Gene Aliases
RGD1561059
Chromosome Location
Chr. 3: 57257394 - 57646666 on Build Rnor_6.0
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq XM_008761954.1 XP_008760176.1 4 706
XM_008761955.1 XP_008760177.1 4 595

View Full Product Details