Gene Transcripts

Gene Symbol
Vom2r77
Entrez Gene ID
690227
Gene Name
vomeronasal 2 receptor, 77
Gene Aliases
Chromosome Location
Chr. 9: 7749090 - 7815144 on Build Rnor_6.0
UniGene ID
Rn.217642
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001099519.1 NP_001092989.1 6 1782

View Full Product Details