Gene Transcripts

Gene Symbol
Myl6
Entrez Gene ID
685867
Gene Name
myosin, light chain 6, alkali, smooth muscle and non-muscle
Gene Aliases
Chromosome Location
Chr. 7: 2906137 - 2909179 on Build Rnor_6.0
UniGene ID
Rn.146093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109484.1 NP_001102954.1 4 331
XM_008765039.1 XP_008763261.1 4 325
XM_008765040.1 XP_008763262.1 4 325
GenBank mRNA BC166586.1 - 4 296
FQ217612.1 - 4 292
FQ217952.1 - 4 304
FQ221192.1 - 4 298
FQ221499.1 - 4 300
FQ221709.1 - 4 332
FQ221718.1 - 4 303
FQ221751.1 - 4 299
FQ221963.1 - 4 300
FQ222034.1 - 4 291
FQ222160.1 - 4 301
FQ222384.1 - 4 303
FQ222504.1 - 4 280
FQ223317.1 - 4 306
FQ228533.1 - 4 303

Gene Symbol
Myl6b
Entrez Gene ID
685883
Gene Name
myosin, light chain 6B, alkali, smooth muscle and non-muscle
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.146093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109486.1 NP_001102956.1 4 331

Gene Symbol
Myl6l
Entrez Gene ID
362816
Gene Name
myosin, light polypeptide 6, alkali, smooth muscle and non-muscle-like
Gene Aliases
LC17, MLC-3, Myl6
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.146093
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001100983.1 NP_001094453.1 3 275
GenBank mRNA S77858.1 - 3 275

View Full Product Details