Gene Transcripts

Gene Symbol
Zbtb32
Entrez Gene ID
688845
Gene Name
zinc finger and BTB domain containing 32
Gene Aliases
Chromosome Location
Chr. 1: 89043121 - 89047098 on Build Rnor_6.0
UniGene ID
Rn.218711
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001109517.1 NP_001102987.1 1, 2 911
XM_006228793.2 XP_006228855.1 2, 3 1583
XM_008759188.1 XP_008757410.1 2, 3 1936

View Full Product Details