Gene Transcripts

Gene Symbol
Larp7
Entrez Gene ID
686883
Gene Name
La ribonucleoprotein domain family, member 7
Gene Aliases
Chromosome Location
Chr. 2: 231866888 - 231882002 on Build Rnor_6.0
UniGene ID
Rn.79854
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001044290.2 NP_001037755.2 12 1691
GenBank mRNA BC083898.1 - 12 1628

View Full Product Details