Gene Transcripts

Gene Symbol
Tomm40l
Entrez Gene ID
304971
Gene Name
translocase of outer mitochondrial membrane 40 homolog (yeast)-like
Gene Aliases
RGD1562006, Tomm40b
Chromosome Location
Chr. 13: 89590076 - 89595308 on Build Rnor_6.0
UniGene ID
Rn.113744
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001083338.1 NP_001076807.1 5, 6 490
XM_006250247.2 XP_006250309.1 6, 7 969
GenBank mRNA AB274732.1 - 5, 6 490

View Full Product Details