Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_007531.2 | NP_031557.2 | 8 | 986 | |
GenBank mRNA | AK002334.1 | - | 8 | 952 | |
AK011107.1 | - | 8 | 953 | ||
AK090088.1 | - | 8 | 943 | ||
AK132052.1 | - | 8 | 948 | ||
AK132058.1 | - | 8 | 975 | ||
AK135729.1 | - | 8 | 903 | ||
AK151952.1 | - | 8 | 980 | ||
AK152375.1 | - | 7 | 1635 | ||
AK153368.1 | - | 7 | 1635 | ||
AY211613.1 | - | 8 | 948 | ||
AY319418.1 | - | 8 | 838 | ||
BC145875.1 | - | 8 | 938 | 40131178 | |
X78683.1 | - | 8 | 942 |