Gene Transcripts
- Gene Symbol
- Igkv6-23
- Entrez Gene ID
- 637227
- Gene Name
- immunoglobulin kappa variable 6-23
- Gene Aliases
- 19-23, EG637227, Gm7202, IgK
- Chromosome Location
- Chr. 6: 70260409 - 70260933 on Build GRCm38
- UniGene ID
- Mm.305097
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AF045512.1 |
- |
2 |
299 |
|
BC091754.1 |
- |
|
314 |
|
X78108.1 |
- |
2 |
305 |
|
- Gene Symbol
- Igkv4-53
- Entrez Gene ID
- 546213
- Gene Name
- immunoglobulin kappa variable 4-53
- Gene Aliases
- Gm1420, Gm1526, IgVkkh4, kh4
- Chromosome Location
- Chr. 6: 69648824 - 69649354 on Build GRCm38
- UniGene ID
- Mm.305097
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
M36245.1 |
- |
|
221 |
|
- Gene Symbol
- Igkv6-14
- Entrez Gene ID
- 667881
- Gene Name
- immunoglobulin kappa variable 6-14
- Gene Aliases
- Igk-V19-14
- Chromosome Location
- Chr. 6: 70434952 - 70435476 on Build GRCm38
- UniGene ID
- Mm.426557
- Species
- Mus musculus
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
U59155.1 |
- |
|
239 |
|
View Full Product Details