Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001318872.1 | NP_001305801.1 | 5 | 1342 | |
NM_006567.4 | NP_006558.1 | 5 | 1336 | ||
XM_005248812.3 | XP_005248869.1 | 6 | 1400 | ||
XM_011514247.2 | XP_011512549.1 | 5 | 1311 | ||
XM_011514248.2 | XP_011512550.1 | 5 | 1311 | ||
XM_011514249.1 | XP_011512551.1 | 5 | 1311 | ||
XM_017010186.1 | XP_016865675.1 | 5 | 2119 | ||
XM_017010187.1 | XP_016865676.1 | 6 | 2121 | ||
GenBank mRNA | AF097441.1 | - | 4 | 992 | |
AF161438.1 | - | 4 | 661 | ||
AK223423.1 | - | 5 | 1136 | ||
AK312454.1 | - | 5 | 1316 | ||
BC020239.1 | - | 5 | 1136 | 5258249 | |
BC021112.1 | - | 5 | 1136 | 5088776 | |
CR542279.1 | - | 4 | 980 |