Gene Transcripts

Gene Symbol
Lrrc36
Entrez Gene ID
270091
Gene Name
leucine rich repeat containing 36
Gene Aliases
BC036564
Chromosome Location
Chr. 8: 105411389 - 105464086 on Build GRCm38
UniGene ID
Mm.212709
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001033371.3 NP_001028543.1 5 733
NM_001170788.1 NP_001164259.1 8 1062
NM_001170789.1 NP_001164260.1 5 739
GenBank mRNA AK133042.1 - 5 733
BC036564.1 - 3 361
BC150957.1 - 5 725 9087868
BC172120.1 - 5 731 9054406

View Full Product Details