Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_145070.3 | NP_659507.3 | 12, 13 | 1190 | |
GenBank mRNA | AF221713.1 | - | 12, 13 | 1055 | |
AK014542.1 | - | 12, 13 | 1168 | ||
AK129182.1 | - | 12, 13 | 1146 | ||
AK146663.1 | - | 12, 13 | 1071 | ||
BC036288.1 | - | 6, 7 | 523 | ||
BC062914.1 | - | 12, 13 | 1071 | ||
BC065101.1 | - | 12, 13 | 1179 | 30535741 |