Gene Transcripts
- Gene Symbol
- DCUN1D4
- Entrez Gene ID
- 23142
- Gene Name
- defective in cullin neddylation 1 domain containing 4
- Gene Aliases
- Chromosome Location
- Chr. 4: 51843000 - 51916837 on Build GRCh38
- UniGene ID
- Hs.605388
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_001040402.2 |
NP_001035492.1
|
2, 3 |
347 |
|
NM_001287755.1 |
NP_001274684.1
|
2, 3 |
243 |
|
NM_001287757.1 |
NP_001274686.1
|
3, 4 |
510 |
|
NM_015115.3 |
NP_055930.2
|
2, 3 |
347 |
|
XM_005265731.3 |
XP_005265788.1
|
2, 3 |
237 |
|
XM_011534379.2 |
XP_011532681.1
|
2, 3 |
770 |
|
XM_011534380.1 |
XP_011532682.1
|
2, 3 |
106 |
|
XM_017007912.1 |
XP_016863401.1
|
2, 3 |
200 |
|
XM_017007913.1 |
XP_016863402.1
|
1, 2 |
79 |
|
GenBank mRNA |
AK027051.1 |
- |
2, 3 |
166 |
|
AK294894.1 |
- |
2, 3 |
243 |
|
AK294896.1 |
- |
3, 4 |
357 |
|
BC053897.1 |
- |
2, 3 |
213 |
6173607 |
BX537944.1 |
- |
2, 3 |
174 |
|
D87466.1 |
- |
2, 3 |
157 |
|
DA770685.1 |
- |
2, 3 |
237 |
|
View Full Product Details