Gene Transcripts

Gene Symbol
MRPL50
Entrez Gene ID
54534
Gene Name
mitochondrial ribosomal protein L50
Gene Aliases
MRP-L50
Chromosome Location
Chr. 9: 101389967 - 101398637 on Build GRCh38
UniGene ID
Hs.288224
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_019051.2 NP_061924.1 2 409
GenBank mRNA AK000500.1 - 2 386
BC032008.1 - 2 371 4749199
CN388719.1 - 2 419

View Full Product Details