Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001317177.1 | NP_001304106.1 | 6 | 652 | |
NM_017812.3 | NP_060282.1 | 5 | 637 | ||
NR_133671.1 | 6 | 727 | |||
GenBank mRNA | AK000427.1 | - | 5 | 553 | |
AK129533.1 | - | 5 | 518 | ||
AK310236.1 | - | 6 | 639 | ||
AL516679.3 | - | 6 | 549 | ||
BC011596.2 | - | 5 | 559 | 3346346 | |
BC014839.1 | - | 5 | 561 | 2901125 |