Gene Transcripts
- Gene Symbol
- ATMIN
- Entrez Gene ID
- 23300
- Gene Name
- ATM interactor
- Gene Aliases
- ASCIZ, ZNF822
- Chromosome Location
- Chr. 16: 81035853 - 81047346 on Build GRCh38
- UniGene ID
- Hs.16349
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
AK302556.1 |
- |
3 |
2429 |
|
- Gene Symbol
- LOC105374297
- Entrez Gene ID
- 105374297
- Gene Name
- uncharacterized LOC105374297
- Gene Aliases
- Chromosome Location
- Chr. 3: 195646696 - 195652159 on Build GRCh38
- UniGene ID
- Hs.655175
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
GenBank mRNA |
BC044310.1 |
- |
4 |
672 |
|
View Full Product Details