Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001252100.1 | NP_001239029.1 | 35 | 5148 | |
NM_017596.3 | NP_060066.2 | 34 | 5109 | ||
XM_017000731.1 | XP_016856220.1 | 34 | 4791 | ||
XM_017000732.1 | XP_016856221.1 | 29 | 3932 | ||
GenBank mRNA | AB007918.2 | - | 34 | 5109 | |
AM177180.1 | - | 35 | 4837 |