Gene Transcripts

Gene Symbol
KRTAP9-9
Entrez Gene ID
81870
Gene Name
keratin associated protein 9-9
Gene Aliases
KAP9.5, KAP9.9, KRTAP9-5, KRTAP9.9
Chromosome Location
Chr. 17: 41255384 - 41256364 on Build GRCh38
UniGene ID
Hs.722077
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001318227.1 NP_001305156.1 2 953
NM_030975.2 NP_112237.2 1 938
GenBank mRNA AJ406949.1 - 1 764
AJ406951.1 - 3 893

View Full Product Details