Gene Transcripts

Gene Symbol
Cox6b1
Entrez Gene ID
110323
Gene Name
cytochrome c oxidase, subunit VIb polypeptide 1
Gene Aliases
2010000G05Rik, Cox6b
Chromosome Location
Chr. 7: 30616974 - 30626151 on Build GRCm38
UniGene ID
Mm.400
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_025628.2 NP_079904.1 4 344
GenBank mRNA AK007996.1 - 4 345
AK012686.1 - 4 310
AK018721.1 - 4 279
BC024343.1 - 4 287 4989436

View Full Product Details