Gene Transcripts
- Gene Symbol
- Vmn2r48
- Entrez Gene ID
- 625580
- Gene Name
- vomeronasal 2, receptor 48
- Gene Aliases
- EG625580
- Chromosome Location
- Chr. 7: 9927624 - 9953585 on Build GRCm38
- UniGene ID
- Mm.469893
- Species
- Mus musculus
- Gene Symbol
- Vmn2r40
- Entrez Gene ID
- 100042781
- Gene Name
- vomeronasal 2, receptor 40
- Gene Aliases
- Chromosome Location
- Chr. 7: 8907734 - 8931402 on Build GRCm38
- UniGene ID
- Mm.469893
- Species
- Mus musculus
- Gene Symbol
- Vmn2r41
- Entrez Gene ID
- 100042848
- Gene Name
- vomeronasal 2, receptor 41
- Gene Aliases
- Chromosome Location
- Chr. 7: 8137905 - 8161551 on Build GRCm38
- UniGene ID
- Mm.469892
- Species
- Mus musculus
View Full Product Details