Gene Transcripts

Gene Symbol
Dnajc3
Entrez Gene ID
100037258
Gene Name
DnaJ (Hsp40) homolog, subfamily C, member 3
Gene Aliases
AA408985, AU067833, Dnajc3a, Dnajc3b, Prkri, mp58, p58, p58IPK
Chromosome Location
Chr. 14: 118937932 - 118981702 on Build GRCm38
UniGene ID
Mm.12616
Species
Mus musculus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_008929.3 NP_032955.2 5 669
GenBank mRNA AK146097.1 - 5 633
AK148270.1 - 5 626
BC013766.1 - 5 614 2646147
U28423.1 - 5 612

View Full Product Details