Gene Transcripts

Gene Symbol
RT1-CE11
Entrez Gene ID
414791
Gene Name
RT1 class I, locus CE11
Gene Aliases
Chromosome Location
Chr. : - on Build
UniGene ID
Rn.233729
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001008834.1 NP_001008834.1 2 87

View Full Product Details