Gene Transcripts

Gene Symbol
Plac9
Entrez Gene ID
361105
Gene Name
placenta-specific 9
Gene Aliases
Chromosome Location
Chr. 16: 3851102 - 3866008 on Build Rnor_6.0
UniGene ID
Rn.145159
Species
Rattus norvegicus
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001108395.1 NP_001101865.1 3 233
XM_006252630.2 XP_006252692.1 3 482

View Full Product Details