Gene Transcripts

Gene Symbol
ICMT
Entrez Gene ID
23463
Gene Name
isoprenylcysteine carboxyl methyltransferase
Gene Aliases
HSTE14, MST098, MSTP098, PCCMT, PCMT, PPMT
Chromosome Location
Chr. 1: 6221193 - 6235984 on Build GRCh38
UniGene ID
Hs.515688
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_012405.3 NP_036537.1 3 465
XM_011541140.2 XP_011539442.1 2 165
GenBank mRNA AF064084.1 - 3 435
AK092890.1 - 4 406
BC017037.1 - 4 560
BC028168.1 - 3 421 5245125
CR541671.1 - 3 392
CR541711.1 - 3 392

View Full Product Details