Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001320227.1 | NP_001307156.1 | 3 | 296 | |
NM_015938.4 | NP_057022.2 | 3 | 296 | ||
XM_005247511.1 | XP_005247568.1 | 3 | 246 | ||
XM_005247512.1 | XP_005247569.1 | 3 | 577 | ||
GenBank mRNA | AF132941.1 | - | 3 | 267 | |
AK094749.1 | - | 2 | 599 | ||
AK225664.1 | - | 3 | 278 | ||
AK308865.1 | - | 3 | 275 | ||
BC013317.1 | - | 3 | 279 | 4291797 | |
DB480542.1 | - | 3 | 299 |