Gene Transcripts
- Gene Symbol
- VPS28
- Entrez Gene ID
- 51160
- Gene Name
- VPS28, ESCRT-I subunit
- Gene Aliases
- Chromosome Location
- Chr. 8: 144423601 - 144428563 on Build GRCh38
- UniGene ID
- Hs.418175
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NM_016208.3 |
NP_057292.1
|
3, 4 |
184 |
|
NM_183057.2 |
NP_898880.1
|
3, 4 |
184 |
|
XM_005272323.3 |
XP_005272380.1
|
3, 4 |
176 |
|
XM_005272324.3 |
XP_005272381.1
|
3, 4 |
176 |
|
XM_005272325.3 |
XP_005272382.1
|
3, 4 |
169 |
|
XM_011517099.1 |
XP_011515401.1
|
3, 4 |
134 |
|
XM_011517100.1 |
XP_011515402.1
|
3, 4 |
134 |
|
XM_017013529.1 |
XP_016869018.1
|
3, 4 |
169 |
|
GenBank mRNA |
AF182844.1 |
- |
2, 3 |
136 |
|
AF316887.1 |
- |
2, 3 |
118 |
|
AK026865.1 |
- |
3, 4 |
134 |
|
AK090942.1 |
- |
3, 4 |
134 |
|
AK091979.1 |
- |
3 |
250 |
|
AK310062.1 |
- |
3 |
292 |
|
BC006485.1 |
- |
2, 3 |
120 |
3535787 |
BC019321.1 |
- |
2, 3 |
120 |
2822758 |
BC050713.2 |
- |
3, 4 |
132 |
5741129 |
BP380505.1 |
- |
3, 4 |
187 |
|
BU658837.1 |
- |
3, 4 |
477 |
|
KU178635.1 |
- |
2, 3 |
75 |
|
KU178636.1 |
- |
2, 3 |
75 |
|
R72904.1 |
- |
2, 3 |
66 |
|
View Full Product Details