Gene Transcripts

Gene Symbol
SYTL2
Entrez Gene ID
54843
Gene Name
synaptotagmin like 2
Gene Aliases
CHR11SYT, EXO4, PPP1R151, SGA72M, SLP2, SLP2A
Chromosome Location
Chr. 11: 85694221 - 85811182 on Build GRCh38
UniGene ID
Hs.369520
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001162951.2 NP_001156423.1 18 3024
NM_001162952.2 NP_001156424.1 10 1554
NM_001162953.2 NP_001156425.1 18 3027
NM_001289608.1 NP_001276537.1 19 2833
NM_001289609.1 NP_001276538.1 11 1667
NM_001289610.1 NP_001276539.1 11 1238
NM_032943.4 NP_116561.1 19 3203
NM_206929.3 NP_996812.1 12 1358
NM_206930.3 NP_996813.1 9 1531
XM_005274057.1 XP_005274114.1 18 6663
XM_005274060.1 XP_005274117.1 19 2847
XM_005274061.1 XP_005274118.1 19 2844
XM_005274062.2 XP_005274119.1 20 3268
XM_005274063.1 XP_005274120.1 18 2727
XM_005274066.1 XP_005274123.1 17 2676
XM_005274067.4 XP_005274124.1 11 2970
XM_011545105.1 XP_011543407.1 19 6711
XM_011545106.1 XP_011543408.1 19 6708
XM_011545107.2 XP_011543409.1 20 7132
XM_011545108.1 XP_011543410.1 18 6591
XM_011545109.2 XP_011543411.1 19 6693
XM_011545110.1 XP_011543412.1 18 6540
XM_011545114.2 XP_011543416.1 10 2876
XM_011545115.1 XP_011543417.1 8 970
XM_011545116.2 XP_011543418.1 9 2755
XM_017017933.1 XP_016873422.1 18 6660
XM_017017934.1 XP_016873423.1 20 7129
XM_017017935.1 XP_016873424.1 16 6507
XM_017017936.1 XP_016873425.1 16 6205
XM_017017937.1 XP_016873426.1 19 2748
XM_017017938.1 XP_016873427.1 18 2706
XM_017017939.1 XP_016873428.1 12 3014
XM_017017940.1 XP_016873429.1 11 2966
GenBank mRNA AB046817.1 - 19 3214
AK024872.1 - 9 1695
AK074737.1 - 12 1315
AK092121.1 - 9 1349
AK124754.1 - 12 2939
AK127504.1 - 13 3901
AK131365.1 - 12 1831
AK222707.1 - 9 1695
AK222710.1 - 9 1695
AK296224.1 - 19 2815
AK296740.1 - 16 2763
AK298604.1 - 11 1156
AL834422.1 - 6 593
AY386362.1 - 12 5218
BC015540.2 - 9 1025
BC068495.1 - 11 1390
BC110315.1 - 10 1083
BC136450.1 - 18 2959 9020439
BC144114.1 - 18 2959 9052634
BC144115.1 - 18 2956 9052635
BX647671.1 - 13 3654
BX648068.1 - 12 3100
EU035829.1 - 18 2680

View Full Product Details