Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001202554.1 | NP_001189483.1 | 3 | 1621 | |
NM_017837.3 | NP_060307.2 | 3 | 1295 | ||
GenBank mRNA | AK000484.1 | - | 3 | 1070 | |
AK056539.1 | - | 3 | 1614 | ||
AK225521.1 | - | 3 | 1070 | ||
AK301626.1 | - | 4 | 600 | ||
BC013568.1 | - | 3 | 1263 | 3890907 |