Gene Transcripts

Gene Symbol
OTOR
Entrez Gene ID
56914
Gene Name
otoraplin
Gene Aliases
FDP, MIAL1
Chromosome Location
Chr. 20: 16748353 - 16752164 on Build GRCh38
UniGene ID
Hs.41119
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_020157.3 NP_064542.1 2 215
XM_017027959.1 XP_016883448.1 2 191
GenBank mRNA AF233261.1 - 2 210
AF243505.1 - 2 173
AJ242552.1 - 2 166
AY359082.1 - 2 203
BC101688.1 - 2 215 8069194
BC101690.1 - 2 215 8069196

View Full Product Details