Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001850.4 | NP_001841.2 | 5 | 2116 | |
NM_020351.3 | NP_065084.2 | 4 | 2054 | ||
GenBank mRNA | AY550974.1 | - | 3 | 1917 | |
BC013581.1 | - | 5 | 2094 | 3875911 | |
BT009917.1 | - | 2 | 1857 | ||
X57527.1 | - | 2 | 1857 |