Gene Transcripts

Gene Symbol
COX6B1
Entrez Gene ID
1340
Gene Name
cytochrome c oxidase subunit 6B1
Gene Aliases
COX6B, COXG, COXVIb1
Chromosome Location
Chr. 19: 35648223 - 35658784 on Build GRCh38
UniGene ID
Hs.431668
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001863.4 NP_001854.1 4 548
GenBank mRNA BC001015.2 - 4 518
BC002478.2 - 4 417 3347455
X13923.1 - 4 400
X54473.1 - 4 403

View Full Product Details