Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001199687.2 | NP_001186616.1 | 3 | 249 | |
NM_001242310.1 | NP_001229239.1 | 2 | 151 | ||
NM_001324201.1 | NP_001311130.1 | 3 | 249 | ||
NM_001324206.1 | NP_001311135.1 | 3 | 251 | ||
NM_021783.4 | NP_068555.1 | 3 | 251 | ||
NR_136726.1 | 3 | 249 | |||
NR_136727.1 | 3 | 251 | |||
XM_011531000.1 | XP_011529302.1 | 3 | 201 | ||
XM_011531001.1 | XP_011529303.1 | 3 | 249 | ||
XM_011531002.1 | XP_011529304.1 | 3 | 201 | ||
XM_011531003.2 | XP_011529305.1 | 3 | 241 | ||
XM_011531004.1 | XP_011529306.1 | 3 | 201 | ||
XM_011531006.2 | XP_011529308.1 | 3 | 241 | ||
XM_011531007.2 | XP_011529309.1 | 3 | 241 | ||
XM_017029703.1 | XP_016885192.1 | 3 | 241 | ||
XM_017029704.1 | XP_016885193.1 | 3 | 201 | ||
XM_017029706.1 | XP_016885195.1 | 3 | 201 | ||
GenBank mRNA | AF298812.1 | - | 2 | 141 | |
AK296264.1 | - | 3 | 201 | ||
AK314881.1 | - | 3 | 217 | ||
AY152724.1 | - | 2 | 141 | ||
AY358735.1 | - | 2 | 144 | ||
AY358736.1 | - | 2 | 144 | ||
BC034919.1 | - | 3 | 198 | 5434545 | |
BP249106.1 | - | 3 | 217 | ||
DA215848.1 | - | 3 | 250 | ||
DA574050.1 | - | 3 | 201 |