Gene Transcripts

Gene Symbol
OSBPL10
Entrez Gene ID
114884
Gene Name
oxysterol binding protein like 10
Gene Aliases
ORP10, OSBP9
Chromosome Location
Chr. 3: 31660825 - 31981850 on Build GRCh38
UniGene ID
Hs.150122
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_001174060.1 NP_001167531.1 6 1669
NM_017784.4 NP_060254.2 7 1861
XM_005264843.3 XP_005264900.1 7 1637
XM_005264844.4 XP_005264901.1 6 1445
XM_005264845.1 XP_005264902.1 2 545
XM_011533326.2 XP_011531628.1 7 1014
XM_017005667.1 XP_016861156.1 7 1527
XM_017005668.1 XP_016861157.1 7 1504
XM_017005669.1 XP_016861158.1 7 1033
XM_017005670.1 XP_016861159.1 4 1716
GenBank mRNA AB209872.1 - 4 496
AF392451.1 - 8 1571
AK000370.1 - 5 737
AK304067.1 - 6 1122
AL833247.1 - 7 964
BC003168.1 - 7 1265 2905981

View Full Product Details