Gene Transcripts
- Gene Symbol
- TXNDC5
- Entrez Gene ID
- 81567
- Gene Name
- thioredoxin domain containing 5
- Gene Aliases
- ENDOPDI, ERP46, HCC-2, HCC2, PDIA15, STRF8, UNQ364
- Chromosome Location
- Chr. 6: 7881250 - 7910814 on Build GRCh38
- UniGene ID
- Hs.150837
- Species
- Homo sapiens
- Gene Symbol
- BLOC1S5-TXNDC5
- Entrez Gene ID
- 100526836
- Gene Name
- BLOC1S5-TXNDC5 readthrough (NMD candidate)
- Gene Aliases
- MUTED-TXNDC5
- Chromosome Location
- Chr. 6: 7881250 - 8064414 on Build GRCh38
- UniGene ID
- Hs.150837
- Species
- Homo sapiens
Interrogated Sequence |
Translated Protein |
Targeted Exon(s) |
siRNA Location |
IMAGE Clone ID |
RefSeq |
NR_037616.1 |
|
11 |
1157 |
|
GenBank mRNA |
BC052310.1 |
- |
11 |
1107 |
|
View Full Product Details