Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | |
---|---|---|---|---|---|
RefSeq | NM_001014449.2 | NP_001014449.1 | 5 | 620 | |
NM_001014451.2 | NP_001014451.1 | 6 | 630 | ||
NM_001257172.1 | NP_001244101.1 | 6 | 555 | ||
NM_001257173.1 | NP_001244102.1 | 6 | 712 | ||
NM_001257174.1 | NP_001244103.1 | 5 | 666 | ||
NM_001257175.1 | NP_001244104.1 | 4 | 527 | ||
NM_007242.5 | NP_009173.1 | 7 | 723 | ||
XM_006721127.2 | XP_006721190.1 | 6 | 645 | ||
XM_011522832.2 | XP_011521134.1 | 8 | 779 | ||
XM_011522833.2 | XP_011521135.1 | 7 | 686 | ||
XM_011522834.2 | XP_011521136.1 | 7 | 668 | ||
XM_017022890.1 | XP_016878379.1 | 7 | 761 | ||
GenBank mRNA | AF353720.1 | - | 5 | 666 | |
AJ237946.1 | - | 7 | 584 | ||
AK027378.1 | - | 7 | 688 | ||
AK223115.1 | - | 7 | 694 | ||
AK296868.1 | - | 4 | 524 | ||
AK301938.1 | - | 6 | 712 | ||
AK302107.1 | - | 6 | 555 | ||
AK316346.1 | - | 5 | 601 | ||
AL136639.1 | - | 6 | 599 | ||
BC003626.2 | - | 7 | 687 | 3030038 | |
BC010008.2 | - | 5 | 572 | 4298615 | |
BC110805.1 | - | 6 | 494 | ||
BI756088.1 | - | 3 | 381 | ||
CR457215.1 | - | 7 | 584 | ||
CR533509.1 | - | 6 | 491 |