Gene Transcripts

Gene Symbol
DOCK3
Entrez Gene ID
1795
Gene Name
dedicator of cytokinesis 3
Gene Aliases
MOCA, PBP
Chromosome Location
Chr. 3: 50674969 - 51384198 on Build GRCh38
UniGene ID
Hs.476284
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_004947.4 NP_004938.1 15 1292
XM_005264914.3 XP_005264971.1 15 1564
XM_005264915.3 XP_005264972.1 15 1564
XM_005264916.3 XP_005264973.1 15 1342
XM_005264917.3 XP_005264974.1 15 1564
XM_005264918.3 XP_005264975.1 15 1341
XM_006713008.3 XP_006713071.1 15 1564
XM_006713009.3 XP_006713072.1 15 1564
XM_006713010.3 XP_006713073.1 15 1327
XM_011533441.2 XP_011531743.1 4 2173
XM_017005825.1 XP_016861314.1 15 1343
XM_017005826.1 XP_016861315.1 15 1333
XM_017005827.1 XP_016861316.1 6 484
GenBank mRNA AB002297.1 - 10 900
AY145303.1 - 15 1269
AY254099.1 - 15 1292

View Full Product Details