Gene Transcripts

Gene Symbol
KLHDC1
Entrez Gene ID
122773
Gene Name
kelch domain containing 1
Gene Aliases
MST025
Chromosome Location
Chr. 14: 49693105 - 49753152 on Build GRCh38
UniGene ID
Hs.509258
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_172193.2 NP_751943.1 11 1037
XM_011536422.2 XP_011534724.1 11 960
GenBank mRNA AF111806.1 - 11 975
AK127202.1 - 11 960
AL833437.1 - 6 467
BC031270.1 - 11 1000
BC101595.1 - 11 1037 8069101
BC101597.1 - 11 1037 8069103

View Full Product Details