Gene Transcripts

Gene Symbol
WFDC10A
Entrez Gene ID
140832
Gene Name
WAP four-disulfide core domain 10A
Gene Aliases
C20orf146, WAP10, dJ688G8.3
Chromosome Location
Chr. 20: 45629746 - 45631192 on Build GRCh38
UniGene ID
Hs.199380
Species
Homo sapiens
Interrogated Sequence Translated Protein Targeted Exon(s) siRNA Location IMAGE Clone ID
RefSeq NM_080753.2 NP_542791.1 2 206
XM_017027679.1 XP_016883168.1 2 120
GenBank mRNA AY038182.1 - 2 206
BC131579.1 - 2 172 40112102

View Full Product Details